3-Amino-p-toluic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0920-250G
A0920-25G
TCA0920-250GEA
768.67
CAD
TCA0920-250G
TCA0920-025G
3-Amino-p-toluic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-p-toluic acid
CAS Number: 2458-12-0
MDL Number: MFCD00007742
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 166
Other Product Names / Synonyms: 3-Amino-p-toluic Acid
Formula:
C₈H₉NO₂ Boiling Pt: 363 °C (1013 hPa) Melting Pt: 166 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007742 CAS Number: 2458-12-0 EINECS: 219-543-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...