6-Amino-m-cresol ≥98.0%

Supplier: TCI America

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

A0919-25G A0919-250G
TCA0919-025GEA 232.26 CAD
TCA0919-025G TCA0919-250G
6-Amino-m-cresol ≥98.0%
6-Amino-m-cresol

CAS Number: 2835-98-5
MDL Number: MFCD00007693
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 2-Amino-5-methylphenol, 2-Hydroxy-4-methylaniline

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 162
MDL Number: MFCD00007693
CAS Number: 2835-98-5
EINECS: 220-620-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d94703c054a9437f122d9088d5f5777a4826596&vid=3d94703c054a9437f122d9088d5f5777a4826596&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR