Furfuryl acetate ≥97.0%

Supplier: TCI America

A0903-25ML A0903-100ML
TCA0903-025MLEA 47.4 CAD
TCA0903-025ML TCA0903-100ML
Furfuryl acetate ≥97.0%
Furfuryl acetate

CAS Number: 623-17-6
MDL Number: MFCD00003251
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 65
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Acetic Acid Furfuryl Ester

Formula: C₇H₈O₃
Boiling Pt: 177 °C
Density: 1.12 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00003251
CAS Number: 623-17-6
EINECS: 210-775-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe051d5dad9c16d50471d9b5f5a6e943c64df61f&vid=fe051d5dad9c16d50471d9b5f5a6e943c64df61f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR