5-Amino-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0897-5G
A0897-25G
TCA0897-005GEA
104.28
CAD
TCA0897-005G
TCA0897-025G
5-Amino-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
5-Amino-2-nitrobenzoic acid
CAS Number: 13280-60-9
MDL Number: MFCD00035748
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₇H₆N₂O₄ Boiling Pt: 481 °C (1013 hPa) Melting Pt: 234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035748 CAS Number: 13280-60-9 EINECS: 236-283-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...