4-Amino-2-chloropyridine

Supplier: TCI America
Warning

Synonyms: 2-Chloro-4-aminopyridine

A0895-25G A0895-1G A0895-5G
TCA0895-25GEA 224.83 CAD
TCA0895-25G TCA0895-001G TCA0895-005G
4-Amino-2-chloropyridine
4-Amino-2-chloropyridine

CAS Number: 14432-12-3
MDL Number: MFCD00060089
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Form: Crystal
Color: White
Melting point (°C): 92

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 300
Melting Pt: 92 °C
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5674b8c9b80df1c7c4f9ca0840e4356059464240&vid=5674b8c9b80df1c7c4f9ca0840e4356059464240&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR