2-Acetyl-1H-pyrrole ≥98.0%

Supplier: TCI America

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

A0894-5G A0894-25G
TCA0894-005GEA 45.01 CAD
TCA0894-005G TCA0894-025G
2-Acetyl-1H-pyrrole ≥98.0%
2-Acetyl-1H-pyrrole

CAS Number: 1072-83-9
MDL Number: MFCD00005220
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 90

Formula: C₆H₇NO
Boiling Pt: 220 °C
Melting Pt: 90 °C
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8e45af97d63d1cf2199f83d7a459b27e2d0d974&vid=c8e45af97d63d1cf2199f83d7a459b27e2d0d974&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR