4-Acetoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0885-500G A0885-25G
TCA0885-500GEA 368.61 CAD
TCA0885-500G TCA0885-025G
4-Acetoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Acetoxybenzoic acid

CAS Number: 2345-34-8
MDL Number: MFCD00002540
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 193


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Acetic Acid 4-Carboxyphenyl Ester, 4-Carboxyphenyl Acetate

Formula: C₉H₈O₄
Melting Pt: 193 °C
MDL Number: MFCD00002540
CAS Number: 2345-34-8
EINECS: 219-066-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbde010130fbb872234841351dfd4c613562281&vid=bdbde010130fbb872234841351dfd4c613562281&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR