2-Acetylcyclohexanone ≥97.0%

Supplier: TCI America

Synonyms: 2-Acetyl-1-cyclohexanone

A0881-10ML A0881-25ML
TCA0881-010MLEA 52.14 CAD
TCA0881-010ML TCA0881-25ML
2-Acetylcyclohexanone ≥97.0%
2-Acetylcyclohexanone

CAS Number: 874-23-7
MDL Number: MFCD00001633
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 79
Specific Gravity (20/20): 1.06

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 111 °C
Density: 1.06 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001633
CAS Number: 874-23-7
EINECS: 212-858-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=939ce7fc3ddd1ee68853576819f4b0b257d17651&vid=939ce7fc3ddd1ee68853576819f4b0b257d17651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR