3-Amino-4-hydroxybenzoic acid 97,0
Supplier: TCI America
|
Warning
|
A0859-25G
A0859-5G
TCA0859-25GEA
703.1
CAD
TCA0859-25G
TCA0859-005G
3-Amino-4-hydroxybenzoic acid 97,0
3-Amino-4-hydroxybenzoic acid
CAS Number: 1571-72-8
MDL Number: MFCD00007697
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 197
Lambda max.: 308 nm (H2O (pH 11))
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Boiling Pt: 347 Melting Pt: 201 Storage Temperature: Ambient |
MDL Number:
MFCD00007697 CAS Number: 1571-72-8 EINECS: 216-390-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...