2-Aminobenzimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0850-5G A0850-25G
TCA0850-005GEA 54.62 CAD
TCA0850-005G TCA0850-025G
2-Aminobenzimidazole ≥98.0% (by HPLC, titration analysis)
2-Aminobenzimidazole

CAS Number: 934-32-7
MDL Number: MFCD00005596
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 231

Formula: C₇H₇N₃
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 231 °C
MDL Number: MFCD00005596
CAS Number: 934-32-7
EINECS: 213-280-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f32f55e12c418aa5b7b4b046b14bed0b600f087&vid=7f32f55e12c418aa5b7b4b046b14bed0b600f087&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR