3-Acetylindole ≥98.0% (by total nitrogen basis)

Supplier: TCI America

A0849-5G A0849-25G
TCA0849-005GEA 93.25 CAD
TCA0849-005G TCA0849-025G
3-Acetylindole ≥98.0% (by total nitrogen basis)
3-Acetylindole

CAS Number: 703-80-0
MDL Number: MFCD00005626
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 190

Formula: C₁₀H₉NO
Boiling Pt: 144…145 °C (10 mmHg)
Melting Pt: 190 °C
MDL Number: MFCD00005626
CAS Number: 703-80-0
EINECS: 211-875-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c30db85fa5e92eca150d685fec048a703cf32417&vid=c30db85fa5e92eca150d685fec048a703cf32417&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR