N-Aminophthalimide ≥98.0% (by HPLC)

Supplier: TCI America

A0843-5G A0843-25G
TCA0843-5GEA 132.72 CAD
TCA0843-5G TCA0843-025G
N-Aminophthalimide ≥98.0% (by HPLC)
N-Aminophthalimide

CAS Number: 1875-48-5
MDL Number: MFCD00005895
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₈H₆N₂O₂
Boiling Pt: 391 °C (1013 hPa)
Melting Pt: 203…206 °C
MDL Number: MFCD00005895
CAS Number: 1875-48-5
EINECS: 217-505-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1df02721ac60172d6c968c4b3612b0c767520642&vid=1df02721ac60172d6c968c4b3612b0c767520642&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR