2-Amino-5-chloropyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine

A0837-25G A0837-500G A0837-100G
TCA0837-025GEA 42.66 CAD
TCA0837-025G TCA0837-500G TCA0837-100G
2-Amino-5-chloropyridine ≥98.0%
2-Amino-5-chloropyridine

CAS Number: 1072-98-6
MDL Number: MFCD00006324
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 136
Flash Point (°C): 160

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 241
Melting Pt: 136 °C
Density: 1.514 g/cm³ (20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f81c37dd5283893c45a6d532540ebf5af27a25c9&vid=f81c37dd5283893c45a6d532540ebf5af27a25c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR