(S)-(+)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0826-5G A0826-1G
TCA0826-005GEA 134.3 CAD
TCA0826-005G TCA0826-001G
(S)-(+)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)
(S)-(+)-2-Aminobutyric acid

CAS Number: 1492-24-6
MDL Number: MFCD00064415
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 280
Specific rotation [a]20/D: 20 deg (C=5, HCl(2+1))

Formula: C₄H₉NO₂
Melting Pt: 280 °C
MDL Number: MFCD00064415
CAS Number: 1492-24-6
EINECS: 216-083-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e28cb5798ee2357edc4607138873dde809ed57f&vid=0e28cb5798ee2357edc4607138873dde809ed57f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR