tert-Butyl acetoacetate ≥95.0%

Supplier: TCI America

Synonyms: tert-Butyl Acetoacetate

A0816-500ML A0816-25ML A0816-100ML
TCA0816-500MLEA 165.9 CAD
TCA0816-500ML TCA0816-025ML TCA0816-100ML
tert-Butyl acetoacetate ≥95.0%
tert-Butyl acetoacetate

CAS Number: 1694-31-1
MDL Number: MFCD00008811
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 66
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid tert-Butyl Ester

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 190
Melting Pt: -38
Density: 0.97 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00008811
CAS Number: 1694-31-1
EINECS: 216-904-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1468c14f572b5ca843900e594abe7bb3e6048051&vid=1468c14f572b5ca843900e594abe7bb3e6048051&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR