4-Nitro-2-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine

A0805-25G A0805-1G A0805-5G
TCA0805-25GEA 80.58 CAD
TCA0805-25G TCA0805-1G TCA0805-5G
4-Nitro-2-(trifluoromethyl)aniline ≥98.0%
4-Nitro-2-(trifluoromethyl)aniline

CAS Number: 121-01-7
MDL Number: MFCD00007365
Molecular Formula: C7H5F3N2O2
Molecular Weight: 206.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Nitro-2-trifluoromethylaniline

Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 91 °C
Density: 1.754 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&vid=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR