2-Amino-5-nitropyridine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine

A0794-250G A0794-25G
TCA0794-250GEA 660.44 CAD
TCA0794-250G TCA0794-025G
2-Amino-5-nitropyridine ≥98.0%
2-Amino-5-nitropyridine

CAS Number: 4214-76-0
MDL Number: MFCD00006325
Molecular Formula: C5H5N3O2
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 188
Flash Point (°C): 224

Formula: C₅H₅N₃O₂
Melting Pt: 188 °C
Density: 1.623 g/cm³ (20 °C)
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ff28acbedbec533db239cb8553a83aff35deb5&vid=74ff28acbedbec533db239cb8553a83aff35deb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR