2-Aminobenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0788-500G A0788-25G
TCA0788-500GEA 1050.7 CAD
TCA0788-500G TCA0788-025G
2-Aminobenzophenone ≥98.0% (by GC, titration analysis)
2-Aminobenzophenone

CAS Number: 2835-77-0
MDL Number: MFCD00007713
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 108

Formula: C₁₃H₁₁NO
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 108 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e98831d9d33751744d17835619237720398a51f&vid=2e98831d9d33751744d17835619237720398a51f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR