Adamantane-1-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid

A0742-500G A0742-25G A0742-100G
TCA0742-500GEA 676.24 CAD
TCA0742-500G TCA0742-025G TCA0742-100G
Adamantane-1-carboxylic acid ≥98.0%
Adamantane-1-carboxylic acid

CAS Number: 828-51-3
MDL Number: MFCD00074720
Molecular Formula: C11H16O2
Molecular Weight: 180.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 1-Adamantoic Acid

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Melting Pt: 173 °C
Density: 1.221 g/cm³ (20 °C)
MDL Number: MFCD00074720
CAS Number: 828-51-3
EINECS: 212-584-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26148fdb8e37762c773d990dd0918116e5808e9e&vid=26148fdb8e37762c773d990dd0918116e5808e9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR