2-Adamantanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol

A0720-5G A0720-25G
TCA0720-005GEA 67.94 CAD
TCA0720-005G TCA0720-025G
2-Adamantanol ≥98.0%
2-Adamantanol

CAS Number: 700-57-2
MDL Number: MFCD00074744
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 2-Hydroxyadamantane

Formula: C₁₀H₁₆O
Melting Pt: 299…303 °C
Density: 1.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074744
CAS Number: 700-57-2
EINECS: 211-846-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b353b76d8fe856e16e484bc091a18c5688183a19&vid=b353b76d8fe856e16e484bc091a18c5688183a19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR