2-Adamantanone ≥98.0%

Supplier: TCI America

A0719-5G A0719-25G A0719-100G
TCA0719-005GEA 69.36 CAD
TCA0719-005G TCA0719-025G TCA0719-100G
2-Adamantanone ≥98.0%
2-Adamantanone

CAS Number: 700-58-3
MDL Number: MFCD00074737
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₁₀H₁₄O
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 43 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac9e45c13df1a4753c4075ab5e09c9507cd6eae&vid=bac9e45c13df1a4753c4075ab5e09c9507cd6eae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR