2-Adamantylamine hydrochloride ≥99.0%
Supplier: TCI America
Synonyms:
2-Aminoadamantane hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-2-ylamine hydrochloride, 2-Adamantanamine hydrochloride, Adamantan-2-amine hydrochloride, 2-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 2-Adamantanammonium chloride
CAS Number: 10523-68-9
MDL Number: MFCD00074743
Molecular Formula: C10H17N
Molecular Weight: 187.71
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 280
Other Product Names / Synonyms: 2-Aminoadamantane Hydrochloride
Formula:
C₁₀H₁₈ClN MW: 187.71 g/mol Melting Pt: 300 |
MDL Number:
MFCD00074743 CAS Number: 10523-68-9 EINECS: 234-074-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...