2-Amino-6-ethoxybenzothiazole ≥97.0% (by titrimetric analysis)
Supplier: TCI America
A0716-500G
A0716-25G
TCA0716-500GEA
636.42
CAD
TCA0716-500G
TCA0716-025G
2-Amino-6-ethoxybenzothiazole ≥97.0% (by titrimetric analysis)
2-Amino-6-ethoxybenzothiazole
CAS Number: 94-45-1
MDL Number: MFCD00005788
Molecular Formula: C9H10N2OS
Molecular Weight: 194.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 162
Formula:
C₉H₁₀N₂OS Boiling Pt: 347 °C (1013 hPa) Melting Pt: 162 °C |
MDL Number:
MFCD00005788 CAS Number: 94-45-1 EINECS: 202-333-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...