2-Amino-6-methylbenzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 6-Methylbenzothiazol-2-ylamine

A0714-25G A0714-100G
TCA0714-025GEA 129.56 CAD
TCA0714-025G TCA0714-100G
2-Amino-6-methylbenzothiazole ≥98.0%
2-Amino-6-methylbenzothiazole

CAS Number: 2536-91-6
MDL Number: MFCD00005789
Molecular Formula: C8H8N2S
Molecular Weight: 164.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₈H₈N₂S
MW: 164.23 g/mol
Boiling Pt: 321
Melting Pt: 142
MDL Number: MFCD00005789
CAS Number: 2536-91-6
EINECS: 219-801-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e78e8b7d19a2afcb9a23b76808334d900b6b6a&vid=15e78e8b7d19a2afcb9a23b76808334d900b6b6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR