4-Pentyloxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 4-(Pentyloxy)benzaldehyde, 4-n-Pentyloxybenzaldehyde, 4-Amyloxybenzaldehyde, 4-n-Amyloxybenzaldehyde

A0707-25ML A0707-500ML
TCA0707-025MLEA 88.48 CAD
TCA0707-025ML TCA0707-500ML
4-Pentyloxybenzaldehyde ≥98.0%
4-Pentyloxybenzaldehyde

CAS Number: 5736-91-4 MDL Number: MFCD00014135 Molecular Formula: C12H16O2 Molecular Weight: 192.26 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 158 Specific Gravity (20/20): 1.02


Form: Clear Liquid


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Pentyloxybenzaldehyde

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 158 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00014135
CAS Number: 5736-91-4
EINECS: 227-250-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c71a0a4d97530cbe4534928d9ce16c0ed6aed36&vid=136f04c8f52f1a09c0dce546615a52ac2c84c635&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR