4-Allylanisole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Allyl-4-methoxybenzene, Estragole

A0702-10G A0702-100G
TCA0702-010GEA 38.58 CAD
TCA0702-010G TCA0702-100G
4-Allylanisole ≥98.0%
4-Allylanisole

CAS Number: 140-67-0
MDL Number: MFCD00008653
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 81
Specific Gravity (20/20): 0.97


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Estragole, 4-Methoxyallylbenzene

Formula: C₁₀H₁₂O
Boiling Pt: 220 °C
Melting Pt: 22.5 °C
Density: 0.97 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00008653
CAS Number: 140-67-0
EINECS: 205-427-8
Merck Index: 13,03740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fbd13ef26bb83b14fdaafbe5bdb1cb25567e47d&vid=6fbd13ef26bb83b14fdaafbe5bdb1cb25567e47d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR