N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(Carbamoylmethyl)-2-aminoethanesulphonic AcidN-(Carbamoylmethyl)taurineN-(Carbamoylmethyl)-2-aminoethanesulfonic AcidACES

A0700-25G
TCA0700-025GEA 80.75 CAD
TCA0700-025G
N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) ≥99.0% (by titrimetric analysis)
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)

[Good's buffer component for biological research]
CAS Number: 7365-82-4
MDL Number: MFCD00008030
Molecular Formula: C4H10N2O4S
Molecular Weight: 182.19
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: ACES, 2-[(Carbamoylmethyl)amino]ethanesulfonic Acid

Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Melting Pt: 270…295 °C
MDL Number: MFCD00008030
CAS Number: 7365-82-4
Merck Index: 13,00037
https://ca.vwr.com/stibo/web/std.lang.all/82/83/28928283.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR