ADA ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid

A0699-25G A0699-500G
TCA0699-025GEA 69.52 CAD
TCA0699-025G TCA0699-500G
ADA ≥98.0% (by titrimetric analysis)
ADA

CAS Number: 26239-55-4
MDL Number: MFCD00008031
Molecular Formula: C6H10N2O5
Molecular Weight: 190.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 201


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: ADA, N-(Carbamoylmethyl)iminodiacetic Acid

Formula: H₂NC(O)CH₂N(CH₂COOH)₂
MW: 190.16 g/mol
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00008031
CAS Number: 26239-55-4
EINECS: 247-530-0
Merck Index: 13,00148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3bec2243f88068cfd875a4c5b8ecd8f74b1daa3&vid=d3bec2243f88068cfd875a4c5b8ecd8f74b1daa3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR