Adamantane ≥99.0%
Supplier: TCI America
A0696-25G
A0696-500G
TCA0696-025GEA
54.51
CAD
TCA0696-025G
TCA0696-500G
Adamantane ≥99.0%
Adamantane
CAS Number: 281-23-2
MDL Number: MFCD00074719
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 87
Other Product Names / Synonyms: Tricyclo[3.3.1.1(3,7)]decane
Formula:
C₁₀H₁₆ Boiling Pt: 171 °C (1013 hPa) Melting Pt: –25 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074719 CAS Number: 281-23-2 EINECS: 206-001-4 Merck Index: 13,00149 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...