N-Acetyl-L-glutamic acid ≥98.0%

Supplier: TCI America

A0693-250G A0693-25G
TCA0693-250GEA 208.56 CAD
TCA0693-250G TCA0693-025G
N-Acetyl-L-glutamic acid ≥98.0%
N-Acetyl-L-glutamic acid

CAS Number: 1188-37-0
MDL Number: MFCD00002802
Molecular Formula: C7H11NO5
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198
Specific rotation [a]20/D: -15 deg (C=1, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Glu-OH

Formula: C₇H₁₁NO₅
MW: 189.17 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00002802
CAS Number: 1188-37-0
EINECS: 214-708-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c1e0de64734960d13938d481e5da3b5ae4a7365&vid=1c1e0de64734960d13938d481e5da3b5ae4a7365&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR