2-Phenylethyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate

A0692-25G A0692-500G
TCA0692-025GEA 36.96 CAD
TCA0692-025G TCA0692-500G
2-Phenylethyl acetate ≥98.0%
2-Phenylethyl acetate

CAS Number: 103-45-7
MDL Number: MFCD00008720
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 224
Flash Point (°C): 110
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Acetic Acid Phenethyl Ester, Phenethyl Acetate, Acetic Acid 2-Phenylethyl Ester

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 224 °C
Melting Pt: –31 °C
Density: 1.04 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008720
CAS Number: 103-45-7
EINECS: 203-113-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec7b270a57ee4a9ec1755eca69d414bd0259de3&vid=dec7b270a57ee4a9ec1755eca69d414bd0259de3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR