9-Anthracenecarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A0690-25G A0690-5G
TCA0690-025GEA 199.08 CAD
TCA0690-025G TCA0690-005G
9-Anthracenecarboxylic acid ≥97.0% (by GC, titration analysis)
9-Anthracenecarboxylic acid

CAS Number: 723-62-6
MDL Number: MFCD00001257
Molecular Formula: C15H10O2
Molecular Weight: 222.24
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 219


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 9-Anthroic Acid, 9-Carboxyanthracene

Formula: C₁₅H₁₀O₂
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 219 °C
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6
EINECS: 211-964-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b11c443f9ac0e6de5874bcd139d30c146e371a&vid=11e7af058c6f4300885797b0af81e2c1b2f9a734&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR