α-Acetyl-γ-butyrolactone ≥99.0%
Supplier: TCI America
A0681-25G
A0681-500G
TCA0681-25GEA
34.76
CAD
TCA0681-25G
TCA0681-500G
α-Acetyl-γ-butyrolactone ≥99.0%
α-Acetyl-γ-butyrolactone
CAS Number: 517-23-7
MDL Number: MFCD00005394
Molecular Formula: C6H8O3
Molecular Weight: 128.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 135
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: alpha-Acetyl-gamma-hydroxybutyric Acid Lactone
Formula:
C₆H₈O₃ Boiling Pt: 108 °C Density: 1.19 (20 °C) Flash Pt: 135 °C |
MDL Number:
MFCD00005394 CAS Number: 517-23-7 EINECS: 208-235-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...