Dipentyl sulfide ≥97.0%
Supplier: TCI America
|
Warning
|
A0673-25G
A0673-5G
TCA0673-25GEA
488.22
CAD
TCA0673-25G
TCA0673-5G
Dipentyl sulfide ≥97.0%
Dipentyl sulphide
CAS Number: 872-10-6
MDL Number: MFCD00027275
Molecular Formula: C10H22S
Molecular Weight: 174.35
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 85
Specific Gravity (20/20): 0.84
Other Product Names / Synonyms: Diamyl Sulfide, Dipentyl Sulfide, Pentyl Sulfide
Formula:
C₁₀H₂₂S Boiling Pt: 175 °C Melting Pt: –51.3 °C Density: 0.84 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00027275 CAS Number: 872-10-6 EINECS: 212-820-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...