5-Chloroanthranilic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A0665-5G A0665-25G
TCA0665-005GEA 48.98 CAD
TCA0665-005G TCA0665-025G
5-Chloroanthranilic acid ≥99.0% (by titrimetric analysis)
5-Chloroanthranilic acid

CAS Number: 635-21-2
MDL Number: MFCD00007838
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 208


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Amino-5-chlorobenzoic Acid

Formula: C₇H₆ClNO₂
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00007838
CAS Number: 635-21-2
EINECS: 211-230-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2bb8dc245a78618e29924bb9e87930002e650c3&vid=c71b3fb0b7240763d6a3ac4f0fcc20ad3ac34a1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR