5-Aminovaleric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0663-25G A0663-5G
TCA0663-25GEA 313.28 CAD
TCA0663-25G TCA0663-5G
5-Aminovaleric acid ≥98.0% (by titrimetric analysis)
5-Aminovaleric acid

CAS Number: 660-88-8
MDL Number: MFCD00008232
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 158


Other Product Names / Synonyms: 5-Aminopentanoic Acid, Homopiperidinic Acid

Formula: C₅H₁₁NO₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 158 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008232
CAS Number: 660-88-8
EINECS: 211-544-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2ec311ad029e16e022faadf41c2dcf0baed3f97&vid=e2ec311ad029e16e022faadf41c2dcf0baed3f97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR