6-Aminouracil ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0658-250G A0658-25G
TCA0658-250GEA 565.64 CAD
TCA0658-250G TCA0658-025G
6-Aminouracil ≥98.0% (by titrimetric analysis)
6-Aminouracil

CAS Number: 873-83-6
MDL Number: MFCD00006071
Molecular Formula: C4H5N3O2
Molecular Weight: 127.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 360


Other Product Names / Synonyms: 4-Amino-2,6-dihydroxypyrimidine, 4-Aminouracil, 6-Amino-2,4-pyrimidinediol

Formula: C₄H₅N₃O₂
Melting Pt: 360 °C
MDL Number: MFCD00006071
CAS Number: 873-83-6
EINECS: 212-854-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45f43f7b2b4ba7489816595b17dd179e2ad91d0c&vid=45f43f7b2b4ba7489816595b17dd179e2ad91d0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR