Methyl acetoacetate ≥99.0%

Supplier: TCI America
Warning

A0650-25G A0650-500G
TCA0650-025GEA 36.99 CAD
TCA0650-025G TCA0650-500G
Methyl acetoacetate ≥99.0%
Methyl acetoacetate

CAS Number: 105-45-3
MDL Number: MFCD00008784
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Melting point (°C): -80
Flash Point (°C): 72
Specific Gravity (20/20): 1.08


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Acetoacetic Acid Methyl Ester, Methyl 3-Oxobutyrate, 3-Oxobutyric Acid Methyl Ester, MAA

Formula: C₅H₈O₃
Boiling Pt: 170 °C
Melting Pt: –80 °C
Density: 1.08 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00008784
CAS Number: 105-45-3
EINECS: 203-299-8
Merck Index: 12,06090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca48a428ccee7d3a29c7ca6d11467a8a0c137172&vid=ca48a428ccee7d3a29c7ca6d11467a8a0c137172&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR