3-Amino-4-chlorobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Amino-4-chlorobenzoic acid, 3-Amino-4-chlorobenzoic Acid

A0635-500G A0635-25G
TCA0635-500GEA 325.64 CAD
TCA0635-500G TCA0635-025G
3-Amino-4-chlorobenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-4-chlorobenzoic acid

CAS Number: 2840-28-0
MDL Number: MFCD00007671
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 216

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330
Melting Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00007671
CAS Number: 2840-28-0
EINECS: 220-635-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7304ed554b9237d352146ce09ac4ef0ad9d681af&vid=4808fdc3898995a7c337a1021be7165f31166c2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR