3-Aminobenzamide ≥98.0%

Supplier: TCI America

Synonyms: m-Aminobenzamide, INO-1001

A0630-5G A0630-25G
TCA0630-005GEA 57.89 CAD
TCA0630-005G TCA0630-025G
3-Aminobenzamide ≥98.0%
3-Aminobenzamide

CAS Number: 3544-24-9
MDL Number: MFCD00007989
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 114

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 341
Melting Pt: 114 °C
MDL Number: MFCD00007989
CAS Number: 3544-24-9
EINECS: 222-586-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33d63c5f7d57a58b0eb36cfecb2c1fb1d0b0909b&vid=33d63c5f7d57a58b0eb36cfecb2c1fb1d0b0909b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR