1-Aminopyridinium iodide ≥98.0%

Supplier: TCI America

Synonyms: 1-Aminopyridinium Iodide, 1-Aminopyridine hydroiodide, 1-Aminopyridine HI

A0629-25G A0629-5G
TCA0629-25GEA 320.74 CAD
TCA0629-25G TCA0629-005G
1-Aminopyridinium iodide ≥98.0%
1-Aminopyridinium iodide

CAS Number: 6295-87-0
MDL Number: MFCD00051893
Molecular Formula: C5H7IN2
Molecular Weight: 222.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₇IN₂
Melting Pt: 161…162 °C
MDL Number: MFCD00051893
CAS Number: 6295-87-0
EINECS: 228-566-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dbacadaeb07d1a0c571cde19ff622d26253ff1d&vid=1dbacadaeb07d1a0c571cde19ff622d26253ff1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR