2-Amino-2-ethyl-1,3-propanediol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0620-25G A0620-500G
TCA0620-025GEA 36.94 CAD
TCA0620-025G TCA0620-500G
2-Amino-2-ethyl-1,3-propanediol ≥98.0% (by titrimetric analysis)
2-Amino-2-ethyl-1,3-propanediol

CAS Number: 115-70-8
MDL Number: MFCD00004680
Molecular Formula: C5H13NO2
Molecular Weight: 119.16
Purity/Analysis Method: >98.0% (T)
Form: Lump
Boiling point (°C): 152
Flash Point (°C): 144
Freezing point (°C): 34

Formula: C₅H₁₃NO₂
Boiling Pt: 152 °C
Melting Pt: 39 °C
Density: 1.099 g/cm³ (20 °C)
Flash Pt: 144 °C
MDL Number: MFCD00004680
CAS Number: 115-70-8
EINECS: 204-101-2
Merck Index: 12,00459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45da71b5180e4a9118851c6e27bd0acd0efd3ffc&vid=45da71b5180e4a9118851c6e27bd0acd0efd3ffc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR