Amantadine hydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

A0588-25G A0588-250G
TCA0588-025GEA 72.32 CAD
TCA0588-025G TCA0588-250G
Amantadine hydrochloride ≥99.0% (by titrimetric analysis)
Amantadine hydrochloride

CAS Number: 665-66-7
MDL Number: MFCD00074723
Molecular Formula: C10H17N
Molecular Weight: 187.71
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 360


Product Categories: Bioscience, Neurotransmitter Agent Ingredients, Antiviral Ingredients
Other Product Names / Synonyms: 1-Aminoadamantane Hydrochloride, Amantadine Hydrochloride

Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Melting Pt: 360 °C
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5f5450ef5ca791049eec5f7d1d136a1f7195123&vid=a5f5450ef5ca791049eec5f7d1d136a1f7195123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR