L(+)-Aspartic acid 99,0

Supplier: TCI America

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

A0546-25G A0546-500G
TCA0546-025GEA 46.28 CAD
TCA0546-025G TCA0546-500G
L(+)-Aspartic acid 99,0
L(+)-Aspartic acid

CAS Number: 56-84-8
MDL Number: MFCD00002616
Molecular Formula: C4H7NO4
Molecular Weight: 133.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-Aminosuccinic Acid, L-Asparaginic Acid, H-Asp-OH

Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 311
Melting Pt: 251
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=803bff47aaf667a2bd3a97876712dd529387367f&vid=803bff47aaf667a2bd3a97876712dd529387367f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR