L(+)-Ascorbic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0537-25G A0537-500G
TCA0537-025GEA 38.55 CAD
TCA0537-025G TCA0537-500G
L(+)-Ascorbic acid ≥99.0% (by titrimetric analysis)
L(+)-Ascorbic acid

CAS Number: 50-81-7
MDL Number: MFCD00064328
Molecular Formula: C6H8O6
Molecular Weight: 176.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 192
Specific rotation [a]20/D: 21 deg (C=10, H2O)


Product Categories: Bioscience, Sugar Acids, Sugars (Others), Regenerative Medicine Research Reagents, Antioxidants, ROS/RNS Detection Reagents, Vitamins
Other Product Names / Synonyms: Vitamin C

Formula: C₆H₈O₆
Boiling Pt: 553 °C (1013 hPa)
Melting Pt: 192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d9d2e49c3c04cd88d80bc6cce7970862481dcca&vid=3d9d2e49c3c04cd88d80bc6cce7970862481dcca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR