L-Arginine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0528-500G
A0528-25G
TCA0528-500GEA
175.22
CAD
TCA0528-500G
TCA0528-025G
L-Arginine hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Arginine hydrochloride
CAS Number: 1119-34-2
MDL Number: MFCD00064550
Molecular Formula: C6H14N4O2
Molecular Weight: 210.66
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: 23 deg (C=8, HCl(1+1))
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, ROS/RNS Generators, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Arg-OH.HCl
Formula:
C₆H₁₅ClN₄O₂ Melting Pt: 235 °C Density: 1.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064550 CAS Number: 1119-34-2 EINECS: 214-275-1 Merck Index: 13,00785 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...