Methyl-o-anisate ≥99.0%

Supplier: TCI America

A0483-25ML A0483-500ML A0483-100ML
TCA0483-025MLEA 55.3 CAD
TCA0483-025ML TCA0483-500ML TCA0483-100ML
Methyl-o-anisate ≥99.0%
Methyl o-anisate

CAS Number: 606-45-1
MDL Number: MFCD00008423
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: o-Anisic Acid Methyl Ester, 2-Methoxybenzoic Acid Methyl Ester, Methyl 2-Methoxybenzoate

Formula: C₉H₁₀O₃
Boiling Pt: 228 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008423
CAS Number: 606-45-1
EINECS: 210-118-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=143c5139b12d08de5e4e00469263ff6e2f228137&vid=143c5139b12d08de5e4e00469263ff6e2f228137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR