p-Anisic acid ≥99.0%

Supplier: TCI America

Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid

A0482-500G A0482-25G
TCA0482-500GEA 151.68 CAD
TCA0482-500G TCA0482-025G
p-Anisic acid ≥99.0%
p-Anisic acid

CAS Number: 100-09-4
MDL Number: MFCD00002542
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 275
Melting point (°C): 184


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 4-Methoxybenzoic Acid

Formula: C₈H₈O₃
Boiling Pt: 275 °C
Melting Pt: 184 °C
Density: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89195d9f48e5a310fc4a0850238230d78b334dd9&vid=8063fb679f508b4b4323a70d4cd4bb1f93ccb5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR