o-Anisic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Methoxybenzoic acid

A0481-25G A0481-500G
TCA0481-025GEA 48.98 CAD
TCA0481-025G TCA0481-500G
o-Anisic acid ≥98.0%
o-Anisic acid

CAS Number: 579-75-9
MDL Number: MFCD00002431
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103


Other Product Names / Synonyms: 2-Methoxybenzoic Acid

Formula: C₈H₈O₃
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 103 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS Number: 579-75-9
EINECS: 209-447-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=014aab838aa1202ae800d224c62c57d751c1dde9&vid=c3412493b429b93e26ec94c5e4e07aa8e4a03668&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR