p-Anisaldehyde ≥99.0%

Supplier: TCI America

Synonyms: 4-Methoxybenzaldehyde

A0480-25ML A0480-500ML
TCA0480-025MLEA 67.78 CAD
TCA0480-025ML TCA0480-500ML
p-Anisaldehyde ≥99.0%
p-Anisaldehyde

CAS Number: 123-11-5
MDL Number: MFCD00003385
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Flash Point (°C): 130
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction), Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Methoxybenzaldehyde

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 248 °C
Melting Pt: 0…2 °C
Density: 1.12 (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adb8d412971e865c194bb07ed8486cb3663b6d2c&vid=adb8d412971e865c194bb07ed8486cb3663b6d2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR